Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgJ Antibody (OTI3B3), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB271016APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271016APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271016APC Supplier Novus Biologicals Supplier No. NBP271016APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgJ Monoclonal specifically detects IgJ in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IgJ
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3B3
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias IGCJJCH, immunoglobulin J chain, immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mupolypeptides
Gene Symbols JCHAIN
Host Species Mouse
Immunogen Full length recombinant protein of human IgJ (NP_653247) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3512.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.