Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGJ Monoclonal Antibody (OTI3C7), TrueMAB™, OriGene
SDP

Catalog No. p-200013063
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0471 30 μL
50-167-4915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0471 Supplier OriGene Supplier No. TA504168S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IGJ Monoclonal antibody specifically detects IGJ in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen IGJ
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI3C7
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene APMAP
Gene Accession No. Q9HDC9
Gene Alias 9530090F24Rik; AI323815; IGCJ; Igj; IgJ chain; immunoglobulin J chain; immunoglobulin J polypeptide, linker protein for immunoglobulin alpha and mu polypeptides; immunoglobulin joining chain; immunoglobulin joining gene (not J region); J chain; Jch; JCHAIN; Joining chain of multimeric IgA and IgM
Host Species Mouse
Immunogen Full length human recombinant protein of human C20orf3 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57136
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.