Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Antibody (PIG45A2), Novus Biologicals™
SDP

Catalog No. NBP261070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261070 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261070 Supplier Novus Biologicals Supplier No. NBP261070
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgM Monoclonal specifically detects IgM in Porcine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen IgM
Applications Flow Cytometry, Immunohistochemistry, Functional Assay
Classification Monoclonal
Clone PIG45A2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000
Formulation Ascites with 0.09% Sodium Azide
Gene Alias AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Gene Symbols IGHM
Host Species Mouse
Immunogen Pig immunoglobulins
Purification Method Unpurified
Quantity 0.1 mg
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3507
Target Species Pig, Goat, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.