Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Antibody (RM109) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB301093 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301093 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301093 Supplier Novus Biologicals Supplier No. NBP325956
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IgM Monoclonal antibody specifically detects IgM in Mouse samples. It is validated for ELISA

Specifications

Antigen IgM
Applications ELISA
Classification Monoclonal
Clone RM109
Conjugate Unconjugated
Dilution ELISA : 0.005 ug/ml - 0.2 ug/ml
Formulation PBS
Gene Alias AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Host Species Rabbit
Immunogen Mouse IgM
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3507
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.