Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgM Antibody (V53P3H2*B2), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP250439R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250439R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250439R Supplier Novus Biologicals Supplier No. NBP250439R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgM Monoclonal specifically detects IgM in Fish samples. It is validated for ELISA.

Specifications

Antigen IgM
Applications ELISA
Classification Monoclonal
Clone V53P3H2*B2
Conjugate DyLight 550
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Gene Symbols IGHM
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3507
Target Species Fish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.