Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ (+)-Igmesine hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
(+)-Igmesine hydrochloride is a selective σ1 receptor ligand (KD = 19.1 nM), with little or no activity at σ2 receptors (IC50 > 1000 nM). Inhibits the NMDA-induced increase in cGMP in a concentration-dependent manner (IC50 value is approximately 100 nM). Weak inhibitor of brain 5-HT uptake in vitro. Exhibits neuroprotective and antidepressant effects.
Specifications
Specifications
Chemical Name or Material | (R)-(+)-N-Cyclopropylmethyl-α-ethyl-N-methyl-α-[(2E)-3-phenyl-2-propenyl)benzenemethanamine hydrochloride |
CAS | 130152-35-1 |
Assay Percent Range | >98% |
Quantity | 10 mg |
Solubility | Soluble to 50mM in DMSO |
Molecular Formula | C23H29N.HCl |
Formula Weight | Observed MW: 355.94 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction