Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF2/CD101 Antibody (307707) [DyLight 594], Novus Biologicals™
SDP

Catalog No. NB372417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372417 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372417 Supplier Novus Biologicals Supplier No. FAB3368M
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IGSF2/CD101 Monoclonal specifically detects IGSF2/CD101 in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen IGSF2/CD101
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 307707
Conjugate DyLight 594
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, leukocyte surface protein, member 2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse IGSF2/CD101, Gln21-Tyr1033, Accession # A8E0Y8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9398
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.