Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ IGSF23 Recombinant Protein Antigen
SDP

Catalog No. NBP233722PE Shop All R&D Systems Products
Click to view available options
Quantity:
0.1mL

Catalog No. NBP233722PE

Supplier: Novus Biologicals™ NBP233722PEP

Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human IGSF23. The IGSF23 Recombinant Protein Antigen is derived from E. coli. The IGSF23 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP2-33722. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

Specifications

Gene ID (Entrez) 147710
Species Human
Purification Method Chromatography
Purity >80%
Concentration 0.5mg/mL
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) Blocking/Neutralizing, Control
Gene Symbol IGSF23
Label Type Unlabeled
Molecular Weight (g/mol) 23kDa
Product Type IGSF23
Quantity 0.1mL
Regulatory Status RUO
Source E.Coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-33722. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogen YMCIATNSKKQLVSEPVTISLPKPIMQPTEAEPMEPDPTLSLSGGSAIGL
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.