Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF4A/SynCAM1/CADM1 Antibody (41N3D3), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP214841W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214841W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP214841W Supplier Novus Biologicals Supplier No. NBP214841AF405
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IGSF4A/SynCAM1/CADM1 Monoclonal specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for Western Blot.

Specifications

Antigen IGSF4A/SynCAM1/CADM1
Applications Western Blot
Classification Monoclonal
Clone 41N3D3
Conjugate Alexa Fluor 405
Dilution Western Blot
Gene Accession No. Q9BY67
Gene Alias BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1
Gene Symbols CADM1
Host Species Rat
Immunogen A portion of amino acids 150-200 of human CADM1 was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23705
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in Human and Primates. 98% in Mouse, Dog, Rabbit, Horse and Chinese Hamster.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.