Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF4A/SynCAM1/CADM1 Antibody (41N3D3) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280786
Description
IGSF4A/SynCAM1/CADM1 Monoclonal specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for Western Blot.Specifications
IGSF4A/SynCAM1/CADM1 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 1-5 ug/ml | |
BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
The amino acid sequence used as immunogen for this antibody is 100% homologous in human and primates. 98% in mouse, dog, rabbit, horse and Chinese hamster. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot | |
41N3D3 | |
Unconjugated | |
PBS | |
CADM1 | |
A portion of amino acids 150-200 of human CADM1 was used as the immunogen for this antibody. | |
0.1 mg | |
Growth and Development, Neuronal Cell Markers, Neuroscience | |
23705 | |
Human, Mouse, Canine, Chinese Hamster, Equine, Primate, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction