Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGSF4A/SynCAM1/CADM1 Antibody (41N3D3) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$501.50
Specifications
Antigen | IGSF4A/SynCAM1/CADM1 |
---|---|
Clone | 41N3D3 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-5 ug/ml |
Classification | Monoclonal |
Description
IGSF4A/SynCAM1/CADM1 Monoclonal specifically detects IGSF4A/SynCAM1/CADM1 in Human samples. It is validated for Western Blot.Specifications
IGSF4A/SynCAM1/CADM1 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
PBS | |
23705 | |
A portion of amino acids 150-200 of human CADM1 was used as the immunogen for this antibody. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
41N3D3 | |
Western Blot 1-5 ug/ml | |
Unconjugated | |
Rat | |
Growth and Development, Neuronal Cell Markers, Neuroscience | |
BL2, cell adhesion molecule 1, IgSF4, IGSF4ADKFZp686F1789, IGSF4immunoglobulin superfamily, member 4D variant 2, Immunoglobulin superfamily member 4, immunoglobulin superfamily, member 4, Necl-2, NECL2MGC149785, nectin-like 2, Nectin-like protein 2, RA175, sgIGSF, Spermatogenic immunoglobulin superfamily, ST17, sTSLC-1, Synaptic cell adhesion molecule, SynCAM, synCAM1, SYNCAMMGC51880, TSLC1, TSLC-1, TSLC1/Nectin-like 2/IGSF4, Tumor suppressor in lung cancer 1SYNCAM1 | |
CADM1 | |
IgG1 κ | |
Protein G purified | |
The amino acid sequence used as immunogen for this antibody is 100% homologous in human and primates. 98% in mouse, dog, rabbit, horse and Chinese hamster. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title