Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF4B/SynCAM3/CADM3 Antibody (035), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB332609 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332609 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332609 Supplier Novus Biologicals Supplier No. NBP290605APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IGSF4B/SynCAM3/CADM3 Monoclonal antibody specifically detects IGSF4B/SynCAM3/CADM3 in Mouse samples. It is validated for ELISA

Specifications

Antigen IGSF4B/SynCAM3/CADM3
Applications ELISA
Classification Monoclonal
Clone 035
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias BIgR, Brain immunoglobulin receptor, cell adhesion molecule 3, IgSF4B, IGSF4BSynCAM3, Immunoglobulin superfamily member 4B, member 4B, NECL-1, NECL1TSLC1-like protein 1, Nectin-like protein 1, Synaptic cell adhesion molecule 3, synCAM3, TSLC1-like 1, TSLL1SYNCAM3
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse IGSF4B/SynCAM3/CADM3 (Accession#: NP_444429.1, Met 1-His 328)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 57863
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.