Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF4C/SynCAM4 Antibody (404619), PerCP, Novus Biologicals™
SDP

Catalog No. NB370755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB370755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB370755 Supplier Novus Biologicals Supplier No. FAB4164C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IGSF4C/SynCAM4 Monoclonal specifically detects IGSF4C/SynCAM4 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot.

Specifications

Antigen IGSF4C/SynCAM4
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 404619
Conjugate PerCP
Dilution Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias cell adhesion molecule 4, IgSF4C, IGSF4Cnectin-like 4, Immunoglobulin superfamily member 4C, immunoglobulin superfamily, member 4C, Necl-4, NECL4synCAM4, Nectin-like protein 4, SYNCAM4, TSLC1-like 2, TSLC1-like protein 2, TSLL2SynCAM4
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human IGSF4C/SynCAM4, Gln25-Tyr323, Accession # Q8NFZ8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199731
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.