Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF8/CD316 Antibody (2587G), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB363289 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363289 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363289 Supplier Novus Biologicals Supplier No. FAB31171J
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IGSF8/CD316 Monoclonal antibody specifically detects IGSF8/CD316 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen IGSF8/CD316
Applications Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 2587G
Conjugate Janelia Fluor 646
Dilution Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD316, CD316 antigen, CD81P3LIR-D1, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRLimmunoglobulin superfamily member 8
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-derived human IGSF8/CD316, Arg28-Thr579, Accession # Q969P0-1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 93185
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.