Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF8/CD316 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB322370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322370 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322370 Supplier Novus Biologicals Supplier No. NBP300128MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IGSF8/CD316 Polyclonal antibody specifically detects IGSF8/CD316 in Human samples. It is validated for ELISA

Specifications

Antigen IGSF8/CD316
Applications ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD316, CD316 antigen, CD81P3LIR-D1, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRLimmunoglobulin superfamily member 8
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human IGSF8/CD316 (Uniprot#: Q969P0-1; Met1-Thr579)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 93185
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.