Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGSF9 Antibody (993107), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB310563 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310563 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310563 Supplier Novus Biologicals Supplier No. FAB9140PECY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IGSF9 Monoclonal specifically detects IGSF9 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IGSF9
Applications Flow Cytometry
Classification Monoclonal
Clone 993107
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias FP18798, immunoglobulin superfamily member 9A, immunoglobulin superfamily, member 9, KIAA1355
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IGSF9, Arg21-Leu730, Accession # Q05BQ1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57549
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.