Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgT Antibody (Z55F8*C3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP261127A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP261127A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP261127A32 Supplier Novus Biologicals Supplier No. NBP261127AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgT Monoclonal specifically detects IgT in Fish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen IgT
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z55F8*C3
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias IgZ, Immunoglobulin IgT
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Fish
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.