Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgT Antibody (Z55F8*C3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261127
Description
IgT Monoclonal specifically detects IgT in Fish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
IgT | |
Monoclonal | |
1.0 mg/mL | |
Western Blot neat, ELISA neat, Immunohistochemistry 1:5 | |
IgZ, Immunoglobulin IgT | |
Ovalbumin-conjugated synthetic peptide | |
0.1 mg | |
Fish | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
Z55F8*C3 | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Mouse | |
Protein A purified | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction