Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgT Antibody (Z55F8*C3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | IgT |
---|---|
Clone | Z55F8*C3 |
Dilution | Western Blot neat, ELISA neat, Immunohistochemistry 1:5 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
IgT Monoclonal specifically detects IgT in Fish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
IgT | |
Western Blot neat, ELISA neat, Immunohistochemistry 1:5 | |
Monoclonal | |
Purified | |
Fish | |
IgZ, Immunoglobulin IgT | |
IgG | |
Protein A purified |
Z55F8*C3 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
PBS with 0.02% Sodium Azide | |
Ovalbumin-conjugated synthetic peptide | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title