Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ikaros/IKZF1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP238242 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP238242 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP238242 Supplier Novus Biologicals Supplier No. NBP238242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ikaros/IKZF1 Polyclonal specifically detects Ikaros/IKZF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Ikaros/IKZF1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Simple Western reported by internal validation, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q13422
Gene Alias DNA-binding protein Ikaros, hIk-1, IK1LyF-1, Ikaros (zinc finger protein), IKAROS family zinc finger 1 (Ikaros), Ikaros family zinc finger protein 1, IKAROSLymphoid transcription factor LyF-1, LYF1PRO0758, zinc finger protein, subfamily 1A, 1 (Ikaros), ZNFN1A1CLL-associated antigen KW-6
Gene Symbols IKZF1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: DADEGQDMSQVSGKESPPVSDTPDEGDEPMPIPEDLSTTSGGQQSSKSDRVVASNVKVETQSDEENGRACEMNGEECAEDLRMLDASGEKMNGSH
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10320
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.