Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ikaros/IKZF1 Rabbit anti-Human, Clone: JB50-38, Novus Biologicals™
SDP

Catalog No. NBP275536 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275536 Supplier Novus Biologicals Supplier No. NBP275536

Rabbit Monoclonal Antibody

Ikaros/IKZF1 Monoclonal specifically detects Ikaros/IKZF1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Ikaros/IKZF1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JB50-38
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias DNA-binding protein Ikaros, hIk-1, IK1LyF-1, Ikaros (zinc finger protein), IKAROS family zinc finger 1 (Ikaros), Ikaros family zinc finger protein 1, IKAROSLymphoid transcription factor LyF-1, LYF1PRO0758, zinc finger protein, subfamily 1A, 1 (Ikaros), ZNFN1A1CLL-associated antigen KW-6
Gene Symbols IKZF1
Host Species Rabbit
Immunogen Recombinant protein within Human Ikaros/IKZF1 aa 261-353/ 519. (SwissProt: Q13422 Human)
Molecular Weight of Antigen 58 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 10320.0
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.