Learn More
Description
Specifications
Specifications
| Antigen | IkB-alpha |
| Applications | CyTOF, Flow Cytometry, Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | 6A920 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 μg/mL, Simple Western 1:20, Flow Cytometry 0.25-1 ug/10^6 cells, Immunohistochemistry 1:20-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1 μg/mL, Immunohistochemistry-Paraffin 1:100, CyTOF-ready, Knockout Validated |
| Gene Accession No. | P25963 |
| Gene Alias | alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor |
| Gene Symbols | NFKBIA |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
