Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkB-alpha, p Ser36 Antibody (39A1413), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB10056724K Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056724K 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056724K Supplier Novus Biologicals Supplier No. NB10056724FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IkB-alpha (p Ser32, p Ser36) Monoclonal specifically detects IkB-alpha (p Ser32, p Ser36) in Bovine, Canine, Human, Mouse, Pig, Rat samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen IkB-alpha (p Ser32, p Ser36)
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 39A1413
Conjugate DyLight 680
Dilution Western Blot
Gene Accession No. P25963
Gene Alias alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
Gene Symbols NFKBIA
Host Species Mouse
Immunogen This monoclonal antibody was raised against a synthetic peptide containing phosphorylated serines at amino acid residues 32 and 36 of human IkBa.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4792
Test Specificity IkBa (39A1413 ) [Ser32/Ser36]
Target Species Bovine, Canine, Human, Mouse, Pig, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.