Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkB-alpha Rabbit anti-Human, Clone: 116, Novus Biologicals™
SDP

Catalog No. NB021549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021549 100 μg
NB021550 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021549 Supplier Novus Biologicals Supplier No. NBP290043100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

IkB-alpha Monoclonal specifically detects IkB-alpha in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen IkB-alpha
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 116
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 ul/mg of lysate
Formulation 0.2 um filtered solution in PBS
Gene Alias alpha, I-kappa-B-alpha, IkappaBalphaNF-kappa-B inhibitor alpha, IKBAikB-alpha, IkB-alpha, MAD3, MAD-3, Major histocompatibility complex enhancer-binding protein MAD3, NFKBI, nuclear factor of kappa light chain gene enhancer in B-cells, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
Host Species Rabbit
Immunogen Recombinant Human IkB-alpha Protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Chaperone Mediated Autophagy (CMA), Diabetes Research, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4792
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.