Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkB-beta, p Ser23 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP254761 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254761 0.1 mg
NBP25476105 0.05 mg
NB413817 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP254761 Supplier Novus Biologicals Supplier No. NBP254761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IkB-beta p Ser23 Polyclonal antibody specifically detects IkB-beta p Ser23 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen IkB-beta (p Ser23)
Applications Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50-1:100
Formulation Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Gene Alias beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta
Gene Symbols NFKBIB
Host Species Rabbit
Immunogen IkB-beta (Phospho-Ser23) antibody was raised against a peptide sequence around phosphorylation site of serine 23 (L-G-S (p) -L-G) derived from Human IkappaB-b
Molecular Weight of Antigen 48 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4793
Test Specificity This antibody detects endogenous level of IkB-beta; only when phosphorylated at serine 23.
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.