Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IkB-beta, p Ser23 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $682.00
Specifications
Concentration | 1 mg/mL |
---|---|
Dilution | Immunohistochemistry 1:50-1:100 |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP254761
![]() |
Novus Biologicals
NBP254761 |
0.1 mg |
Each for $682.00
|
|
|||||
NBP25476105
![]() |
Novus Biologicals
NBP2547610.05MG |
0.05 mg |
Each for $426.00
|
|
|||||
NB413817
![]() |
Novus Biologicals
NBP2547610.025MG |
0.025 mg |
Each for $196.10
|
|
|||||
Description
IkB-beta Polyclonal specifically detects IkB-beta in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
1 mg/mL | |
Immunohistochemistry | |
Unconjugated | |
Human, Mouse, Rat | |
4793 | |
IkB-beta (Phospho-Ser23) antibody was raised against a peptide sequence around phosphorylation site of serine 23 (L-G-S (p) -L-G) derived from Human IkappaB-b | |
Primary |
Immunohistochemistry 1:50-1:100 | |
Polyclonal | |
Rabbit | |
beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta | |
NFKBIB | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title