Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IkB-epsilon Rabbit anti-Human, Clone: JM62-63, Novus Biologicals™
SDP

Catalog No. NBP266903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266903 Supplier Novus Biologicals Supplier No. NBP266903

Rabbit Monoclonal Antibody

IkB-epsilon Monoclonal specifically detects IkB-epsilon in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IkB-epsilon
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JM62-63
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias epsilon, ikappaBepsilon, I-kappa-B-epsilon, IkB-E, IKBENF-kappa-B inhibitor epsilon, ikB-epsilon, NF-kappa-BIE, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
Gene Symbols NFKBIE
Host Species Rabbit
Immunogen Synthetic peptide within Human IkB-epsilon aa 175-224/ 500. (SwissProt: O00221 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4794
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.