Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKB zeta Antibody, Novus Biologicals™
SDP

Catalog No. NB10056390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056390 0.1 mg
NB10056390T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056390 Supplier Novus Biologicals Supplier No. NB10056390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IKB zeta Polyclonal specifically detects IKB zeta in Mouse samples. It is validated for Western Blot.

Specifications

Antigen IKB zeta
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q9BYH8, Q9EST8
Gene Alias FLJ30225, FLJ34463, ikappaBzeta, I-kappa-B-zeta, IkappaB-zeta, ikbzeta, IkB-zeta, IL-1 inducible nuclear ankyrin-repeat protein, INAPIkappa B-zeta variant 3, MAILIKBZikB-zeta, Molecule possessing ankyrin repeats induced by lipopolysaccharide, NF-kappa-B inhibitor zeta, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor
Gene Symbols NFKBIZ
Host Species Rabbit
Immunogen Synthetic peptides corresponding to an amino acid range between 270-320 and 670-720 of mouse IkB zeta protein were used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64332
Test Specificity This antibody is reactive against Mouse. Predicted cross-reactivity based on sequence identity: Mouse (100%), Human, (100%), Rat (100%), Chimp (100%), Bovine (100%), Chicken (93%). Not yet tested in other species.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.