Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKB zeta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP18983525UL
Description
IKB zeta Polyclonal specifically detects IKB zeta in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IKB zeta | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ30225, FLJ34463, ikappaBzeta, I-kappa-B-zeta, IkappaB-zeta, ikbzeta, IkB-zeta, IL-1 inducible nuclear ankyrin-repeat protein, INAPIkappa B-zeta variant 3, MAILIKBZikB-zeta, Molecule possessing ankyrin repeats induced by lipopolysaccharide, NF-kappa-B inhibitor zeta, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
Rabbit | |
Affinity Purified | |
RUO | |
64332 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.1mg/mL | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q9BYH8 | |
NFKBIZ | |
This antibody was developed against Recombinant Protein corresponding to amino acids:QGRRALSYVLARKMNALHMLDIKEHNGQSAFQVAVAANQHLIVQDLVNIGAQVNTTDCWGRTPLHVCAEKGHSQVLQAIQKGAVGSNQFVDLEATNYDGLTPLHCAVIAHNAVVHELQRNQQPHSPEVQELLL | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction