Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKB zeta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
| Antigen | IKB zeta |
|---|---|
| Concentration | 0.1mg/mL |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
IKB zeta Polyclonal specifically detects IKB zeta in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| IKB zeta | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9BYH8 | |
| 64332 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:QGRRALSYVLARKMNALHMLDIKEHNGQSAFQVAVAANQHLIVQDLVNIGAQVNTTDCWGRTPLHVCAEKGHSQVLQAIQKGAVGSNQFVDLEATNYDGLTPLHCAVIAHNAVVHELQRNQQPHSPEVQELLL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.1mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ30225, FLJ34463, ikappaBzeta, I-kappa-B-zeta, IkappaB-zeta, ikbzeta, IkB-zeta, IL-1 inducible nuclear ankyrin-repeat protein, INAPIkappa B-zeta variant 3, MAILIKBZikB-zeta, Molecule possessing ankyrin repeats induced by lipopolysaccharide, NF-kappa-B inhibitor zeta, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor | |
| NFKBIZ | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title