Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKBKAP Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290998 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290998 Supplier Novus Biologicals Supplier No. NBP171918APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IKBKAP Polyclonal antibody specifically detects IKBKAP in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen IKBKAP
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1150 and 1200 of human Inhibitor of Kappa Light Polypeptide Gene Enhancer in B-cells, Kiinase Complex-Associated Protein using the numbering given in entry NP_003631.2 (GeneID 8518).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8518
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.