Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKBKAP Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP121645 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121645 0.1 mg
NB018414 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP121645 Supplier Novus Biologicals Supplier No. NBP121645

Mouse Monoclonal Antibody

V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen V5 Epitope Tag
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone SV5-Pk2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:500, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500
Gene Alias V5 epitope tag
Host Species Mouse
Immunogen A recombinant peptide corresponding to the PK tag fused to a class II - restricted T helper cell epitope.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Epitope Tags, V5 Epitope Tags
Primary or Secondary Primary
Test Specificity NBP1-21645 recognizes a small epitope, termed Pk, present on the P/V proteins of the paramyxovirus, SV5. NBP1-21645 has been used to detect recombinant proteins, some of which include transmembrane and secreted proteins, which have been tagged with this epitope. Usually, a 14 amino acid tag has been added to the recombinant proteins, although a smaller epitope of 9 amino acids (that as a peptide inhibit the binding of the monoclonal antibody to its native protein) has also been successfully used. The 14 amino acid epitope is; gly lys pro ile pro asn pro leu leu gly leu asp ser thr. (The 9 amino acid epitope is underlined).
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.