Learn More
Description
Specifications
Specifications
| Antigen | IKK alpha |
| Applications | Chemotaxis, ChIP Assay, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | 14A231 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/ml, Simple Western 10 ug/ml, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 27705798), Flow Cytometry (Intracellular): 0.25-0.5 ug/10^6 cells, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 27121058), Immunocytochemistry/Immunofluorescence 1:10, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 25133425), Flow (Intracellular) Flow Cytometry: (Intracellular): 0.25-0.5 ug/10^6 cells. Use reported in scientific literature (PMID 24804954), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Gene Accession No. | O15111 |
| Gene Alias | conserved helix-loop-helix ubiquitous kinaseIKBKA, EC 2.7.11, EC 2.7.11.10, IkappaB kinase, I-kappa-B kinase 1, I-kappa-B kinase alpha, IkBKA, IKK1I-kappa-B kinase-alpha, IKK-A, IKKAIkB kinase alpha subunit, IKK-alphainhibitor of nuclear factor kappa-B kinase subunit alpha, NFKBIKAIKK-a kinase, Nuclear factor NFkappaB inhibitor kinase alpha, Nuclear factor NF-kappa-B inhibitor kinase alpha, TCF16, TCF-16, Transcription factor 16 |
| Gene Symbols | CHUK |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
