Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK alpha Antibody (14A231) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$207.00 - $572.50
Specifications
Antigen | IKK alpha |
---|---|
Clone | 14A231 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1 ug/ml, Simple Western 10 ug/ml, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 27705798), Flow Cytometry (Intracellular): 0.25-0.5 ug/10^6 cells, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 27121058), Immunocytochemistry/Immunofluorescence 1:10, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 25133425), Flow (Intracellular) Flow Cytometry: (Intracellular): 0.25-0.5 ug/10^6 cells. Use reported in scientific literature (PMID 24804954), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
Applications | Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056704T
![]() |
Novus Biologicals
NB10056704SS |
0.025 mg |
Each for $207.00
|
|
|||||
NB10056704
![]() |
Novus Biologicals
NB10056704 |
0.1 mg |
Each for $572.50
|
|
|||||
Description
IKK alpha Monoclonal specifically detects IKK alpha in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
IKK alpha | |
1 mg/mL | |
Western Blot, ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, mTOR Pathway, Protein Kinase, Signal Transduction | |
O15111 | |
1147 | |
This antibody was raised against a His-tagged full-length human IKK alpha protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
14A231 | |
Western Blot 1 ug/ml, Simple Western 10 ug/ml, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 27705798), Flow Cytometry (Intracellular): 0.25-0.5 ug/10^6 cells, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 27121058), Immunocytochemistry/Immunofluorescence 1:10, Immunoprecipitation 1-2 ug/ml, Immunohistochemistry-Paraffin 5ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 25133425), Flow (Intracellular) Flow Cytometry: (Intracellular): 0.25-0.5 ug/10^6 cells. Use reported in scientific literature (PMID 24804954), Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Primate | |
conserved helix-loop-helix ubiquitous kinaseIKBKA, EC 2.7.11, EC 2.7.11.10, IkappaB kinase, I-kappa-B kinase 1, I-kappa-B kinase alpha, IkBKA, IKK1I-kappa-B kinase-alpha, IKK-A, IKKAIkB kinase alpha subunit, IKK-alphainhibitor of nuclear factor kappa-B kinase subunit alpha, NFKBIKAIKK-a kinase, Nuclear factor NFkappaB inhibitor kinase alpha, Nuclear factor NF-kappa-B inhibitor kinase alpha, TCF16, TCF-16, Transcription factor 16 | |
CHUK | |
IgG1 κ | |
Protein G purified | |
An 85 kDa band should be observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title