Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK alpha Antibody (14A231), PerCP, Novus Biologicals™
SDP

Catalog No. NBP227409P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227409P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227409P Supplier Novus Biologicals Supplier No. NBP227409PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IKK alpha Monoclonal specifically detects IKK alpha in Human, Mouse, Rat, Primate samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen IKK alpha
Applications Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 14A231
Conjugate PerCP
Dilution Flow Cytometry 1 μL/1 million cells, Flow (Intracellular)
Gene Accession No. O15111
Gene Alias conserved helix-loop-helix ubiquitous kinaseIKBKA, EC 2.7.11, EC 2.7.11.10, IkappaB kinase, I-kappa-B kinase 1, I-kappa-B kinase alpha, IkBKA, IKK1I-kappa-B kinase-alpha, IKK-A, IKKAIkB kinase alpha subunit, IKK-alphainhibitor of nuclear factor kappa-B kinase subunit alpha, NFKBIKAIKK-a kinase, Nuclear factor NFkappaB inhibitor kinase alpha, Nuclear factor NF-kappa-B inhibitor kinase alpha, TCF16, TCF-16, Transcription factor 16
Gene Symbols CHUK
Host Species Mouse
Immunogen This antibody was raised against a His-tagged full-length human IKK alpha protein.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, mTOR Pathway, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1147
Test Specificity An 85 kDa band should be observed.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.