Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK beta Antibody (10A9B6), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB10056513Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056513Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056513Z Supplier Novus Biologicals Supplier No. NB10056513AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IKK beta Monoclonal specifically detects IKK beta in Human, Mouse samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), Gel Super Shift Assays, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.

Specifications

Antigen IKK beta
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining), Gel Supershift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10A9B6
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Intracellular)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O14920
Gene Alias EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta
Gene Symbols IKBKB
Host Species Mouse
Immunogen Full-length human IKKb/IKK2 recombinant protein was used as immunogen (NP_001547).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity, mTOR Pathway, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3551
Test Specificity An approx. 87 kDa band should be observed.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.