Learn More
Description
Specifications
Specifications
| Antigen | IKK beta |
| Applications | EMSA, Flow Cytometry, Flow Cytometry (Intracellular Staining), Gel Supershift, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | 10A9B6 |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 - 5 ug/mL, Simple Western 1:12.5, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24825920), Immunoprecipitation 2 - 5 ug/ 10^6 cells, Immunohistochemistry-Frozen reported by customer review, Gel Super Shift Assays reported in scientific literature (PMID 19662631), Flow (Intracellular) |
| Gene Accession No. | O14920 |
| Gene Alias | EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta |
| Gene Symbols | IKBKB |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
