Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK beta Antibody (10A9B6) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 9 publications

$206.50 - $499.50

Specifications

Antigen IKK beta
Clone 10A9B6
Concentration 1.0 mg/ml
Dilution Western Blot 2 - 5 ug/mL, Simple Western 1:12.5, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24825920), Immunoprecipitation 2 - 5 ug/ 10^6 cells, Immunohistochemistry-Frozen reported by customer review, Gel Super Shift Assays reported in scientific literature (PMID 19662631), Flow (Intracellular)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Gel Shift
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056513T
SDP
View Documents
Novus Biologicals
NB10056513SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056513
SDP
View Documents
Novus Biologicals
NB10056513
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

IKK beta Monoclonal specifically detects IKK beta in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Gel Super Shift Assays, Flow (Intracellular).
Specifications

Specifications

IKK beta
1.0 mg/ml
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Gel Shift
Unconjugated
Mouse
Cancer, Immunology, Innate Immunity, mTOR Pathway, Protein Kinase
O14920
3551
Full-length human IKKb/IKK2 recombinant protein was used as immunogen (NP_001547).
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
10A9B6
Western Blot 2 - 5 ug/mL, Simple Western 1:12.5, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24825920), Immunoprecipitation 2 - 5 ug/ 10^6 cells, Immunohistochemistry-Frozen reported by customer review, Gel Super Shift Assays reported in scientific literature (PMID 19662631), Flow (Intracellular)
Monoclonal
Purified
RUO
Human, Mouse
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta
IKBKB
IgG1 κ
Protein G purified
An approx. 87 kDa band should be observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.