Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK beta Antibody (10AG2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB230069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB230069 0.025 mg
NB10056509 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB230069 Supplier Novus Biologicals Supplier No. NB10056509SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 55 publications

IKK beta Monoclonal specifically detects IKK beta in Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot.

Specifications

Antigen IKK beta
Applications CyTOF, Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot
Classification Monoclonal
Clone 10AG2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 4 ug/mL, Simple Western 1:12.5, Flow Cytometry 0.25 - 0.5 ug/ 10^6 cells, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2-5 ug/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Immunoblotting, CyTOF-ready
Gene Accession No. O14920
Gene Alias EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta
Gene Symbols IKBKB
Host Species Mouse
Immunogen Full-length recombinant human IKK beta protein (NP_001547).
Molecular Weight of Antigen 87 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity, mTOR Pathway, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3551
Test Specificity An approx. 87 kDa band should be observed.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.