Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK beta Antibody (10AG2) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 55 publications

$201.00 - $499.50

Specifications

Antigen IKK beta
Clone 10AG2
Concentration 1.0 mg/mL
Applications Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB230069
SDP
View Documents
Novus Biologicals
NB10056509SS
0.025 mg
Each for $201.00
Only null left
Add to Cart
 
NB10056509
SDP
View Documents
Novus Biologicals
NB10056509
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

IKK beta Monoclonal specifically detects IKK beta in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, CyTOF.
Specifications

Specifications

IKK beta
1.0 mg/mL
Monoclonal
Purified
RUO
O14920
3551
Full-length recombinant human IKK beta protein (NP_001547).
Primary
Store at -20C. Avoid freeze-thaw cycles.
87 kDa
10AG2
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry, Flow Cytometry, Immunoprecipitation
Unconjugated
Mouse
Cancer, Immunology, Innate Immunity, mTOR Pathway, Protein Kinase
EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta
IKBKB
IgG1
Protein G purified
An approx. 87 kDa band should be observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.