Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK beta Antibody, Novus Biologicals™
SDP

Catalog No. NB600477SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600477SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600477SS Supplier Novus Biologicals Supplier No. NB600477SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IKK beta Polyclonal specifically detects IKK beta in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IKK beta
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot
Classification Polyclonal
Concentration 80 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000, Simple Western, ELISA 1:1000-1:5000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:10-1:500, Dot Blot
Formulation Antiserum with 0.01% Sodium Azide
Gene Accession No. O14920, O14920, O14920, O14920
Gene Alias EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase 2, I-kappa-B-kinase beta, IkBKB, IKK2FLJ38368, IKK-B, IKK-betaFLJ36218, IKKBFLJ40509, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta, inhibitor of nuclear factor kappa-B kinase subunit beta, MGC131801, NFKBIKBFLJ33771, Nuclear factor NF-kappa-B inhibitor kinase beta
Gene Symbols IKBKB
Host Species Rabbit
Immunogen IKK beta peptide corresponding to the highly conserved C-terminus region of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH). (Uniprot: O14920)
Molecular Weight of Antigen 86.56 kDa
Purification Method Delipidation and Defibrination
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity, mTOR Pathway, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3551
Test Specificity Anti-IKKb may react non-specifically with other proteins. Control peptide will compete only with the specific reaction of antiserum with the IKKb subunit.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.