Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK epsilon/IKBKE Antibody (OTI3C5), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP271490UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271490UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271490UV Supplier Novus Biologicals Supplier No. NBP271490UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IKK epsilon/IKBKE Monoclonal specifically detects IKK epsilon/IKBKE in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen IKK epsilon/IKBKE
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3C5
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase epsilon, IkBKE, IKK-E, IKKEMGC125297, IKK-epsilon, IKKI, IKK-iMGC125295, Inducible I kappa-B kinase, inducible IkappaB kinase, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon, inhibitor of nuclear factor kappa-B kinase subunit epsilon, KIAA0151IKK-related kinase epsilon, MGC125294
Gene Symbols IKBKE
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 474-716 of human IKK epsilon/IKBKE (NP_054721) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9641.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.