Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK epsilon/IKBKE, p Thr501 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB120084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120084 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120084 Supplier Novus Biologicals Supplier No. NBP177829DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IKK epsilon/IKBKE Polyclonal specifically detects IKK epsilon/IKBKE in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IKK epsilon/IKBKE
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.10, I-kappa-B kinase epsilon, IkBKE, IKK-E, IKKEMGC125297, IKK-epsilon, IKKI, IKK-iMGC125295, Inducible I kappa-B kinase, inducible IkappaB kinase, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon, inhibitor of nuclear factor kappa-B kinase subunit epsilon, KIAA0151IKK-related kinase epsilon, MGC125294
Host Species Rabbit
Immunogen IKK epsilon/IKBKE phospho peptide corresponding to a region of the human protein surrounding pT501 conjugated to KLH. (Uniprot: Q14164)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9641
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.