Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK gamma Antibody (46B844), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB10056532U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056532U 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056532U Supplier Novus Biologicals Supplier No. NB10056532UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IKK gamma Monoclonal specifically detects IKK gamma in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen IKK gamma
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 46B844
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry
Gene Accession No. NP_001093326
Gene Alias FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator
Gene Symbols IKBKG
Host Species Mouse
Immunogen Full-length human IKK gamma protein.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 8517
Test Specificity A 48 kDa band is observed.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.