Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK gamma Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB600121725 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.25 mL
0.5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600121725 0.25 mL
NB018206 0.1 mL
NB6001217 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB600121725 Supplier Novus Biologicals Supplier No. NB60012170.25ML

Rabbit Polyclonal Antibody has been used in 2 publications

MMP-9 Polyclonal specifically detects MMP-9 in Human, Guinea Pig, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen MMP-9
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen
Formulation PBS with BSA and sodium azide
Gene Alias 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
Gene Symbols MMP9
Host Species Rabbit
Immunogen A synthetic peptide from the middle region of human MMP-9
Quantity 0.25 mL
Primary or Secondary Primary
Gene ID (Entrez) 4318
Test Specificity This reacts with the 92 kDa pro form and 86 kDa active form of MMP-9.
Target Species Human, Guinea Pig, Bovine, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.