Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IKK gamma Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018217 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018217 0.025 mg
NB600215 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018217 Supplier Novus Biologicals Supplier No. NB600215SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IKK gamma Polyclonal specifically detects IKK gamma in Human samples. It is validated for Western Blot.

Specifications

Antigen IKK gamma
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator
Gene Symbols IKBKG
Host Species Rabbit
Immunogen N-terminus of the human IKK-gamma protein (proprietary).
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 8517
Test Specificity Reacts with the residues 2-13 [NRHLWKSQLCEM] of the IKK-g protein. Recognizes both human isoforms.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.