Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK gamma Antibody - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2120320.025MG
Description
alpha 2-Macroglobulin Monoclonal specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Western Blot.Specifications
alpha 2-Macroglobulin | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-1000 | |
alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
MM0087-4H44 | |
Unconjugated | |
A 0.2 um filtered solution in PBS with No Preservative | |
A2M | |
Human alpha 2 Macroglobulin | |
0.025 mg | |
Neuroscience | |
2 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction