Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IKK gamma Antibody - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$182.00
Specifications
Antigen | alpha 2-Macroglobulin |
---|---|
Clone | MM0087-4H44 |
Concentration | LYOPH |
Applications | Western Blot |
Classification | Monoclonal |
Description
alpha 2-Macroglobulin Monoclonal specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Western Blot.Specifications
alpha 2-Macroglobulin | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
2 | |
IgG1 | |
Protein G purified |
MM0087-4H44 | |
Western Blot | |
Unconjugated | |
Mouse | |
Neuroscience | |
alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
A2M | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title