Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGFR1/Flt-1 Antibody (EWC) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$243.50 - $560.50
Specifications
| Antigen | VEGFR1/Flt-1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 1-10 ug/ml, ELISA 1.5-8 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, In vivo assay |
| Applications | In vitro Assay, Immunofluorescence, ELISA, Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
Description
VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, In vivo assay.Specifications
| VEGFR1/Flt-1 | |
| Western Blot 1-10 ug/ml, ELISA 1.5-8 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, In vivo assay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1 | |
| FLT1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| In vitro Assay, Immunofluorescence, ELISA, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, HIF Target Genes, Hypoxia, Stem Cell Markers | |
| P17948 | |
| 2321 | |
| IgG1 | |
| Protein G purified | |
| The monoclonal will detect native and denaturated human VEGFR-1/ Flt-1 in ELISA experiments and on the surface of different human cell types. The will also detect mouse Flt-1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title