Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR1/Flt-1 Antibody (EWC) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB6001004 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001004 0.1 mg
NB018197 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6001004 Supplier Novus Biologicals Supplier No. NB6001004
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

VEGFR1/Flt-1 Monoclonal specifically detects VEGFR1/Flt-1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, In vivo assay.

Specifications

Antigen VEGFR1/Flt-1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, In vitro Assay
Classification Monoclonal
Clone EWC
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-10 ug/ml, ELISA 1.5-8 ug/ml, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, In vivo assay
Formulation PBS with No Preservative
Gene Accession No. P17948
Gene Alias EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1
Gene Symbols FLT1
Host Species Mouse
Immunogen Recombinant human soluble extracellular Flt-1 (D5)
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, HIF Target Genes, Hypoxia, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2321
Test Specificity The monoclonal will detect native and denaturated human VEGFR-1/ Flt-1 in ELISA experiments and on the surface of different human cell types. The will also detect mouse Flt-1.
Reconstitution Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. Please note the sample size of this product will be provided in reconstituted liquid form.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.